missing translation for 'onlineSavingsMsg'
Learn More

Hammond™ Drierite™ Desiccant Bags, Cole-Parmer™

Product Code. 11893082 Shop All Cole-Parmer Products
Change view
Click to view available options
Quantity:
3 g
6 g
1 oz.
8 oz.
Unit Size:
Pack of 100
Pack of 200
Pack of 750
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11893082 6 g Pack of 750
11832692 3 g Pack of 750
11803092 1 oz. Pack of 200
11833092 8 oz. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11893082 Supplier Cole Parmer Supplier No. 0719306
  • ONLINE EXCLUSIVE PRICE
    216.00 €
    156.12 € / Pack of 750
    Save 59.88 €
    27% Off
View all promotions
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protect materials from corrosion with these desiccant bags. Hammond™ Drierite™ Desiccant Bags reduce the humidity to a 100°F dew point in 10 hours and are dust-tight. Materials packaged with these bags are protected from corrosion for many months.

Nonwoven crepe bags filled with Drierite (anyhydrous calcium sulfate) completely dry the air within a package or container during shipment or storage.
  • Sturdy and held together by heat seals
  • Perform well in elevated temperatures
  • Will not release absorbed moisture at less than 350°F

Specifications

Material Calcium Sulfate
Quantity 6 g
Suitability These Bags are Dust-Tight and Will Protect For Many Months
compliance-icons
Product Identifier
  • Drierite
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H360F-May damage fertility.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: cobalt dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.