Learn More
Hammond™ Drierite 26800 Drying Column, Cole-Parmer™
Product Code. 11785609 Shop All Cole-Parmer Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity Unit Size
11785609 1 Ea. Each
1 options
Product Code. 11785609 Supplier Cole Parmer Supplier No. 0719300
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Connect the column in-line between a dryer or desiccator and a compressed air line or gas cylinder. Hammond™ Drierite 26800 Drying Column handles a flow of 200 liters per hour or 0.1cfm.
Polycarbonate drying column dries air and gas to a dew point of –100°F at up to 90psi. Column is made up of a plastic cap, 303 stainless steel desiccant supports and coil springs, felt filters, and O-ring gasket. Drying column comes filled with 1.25 lb. of 8 mesh indicating desiccant (anhydrous calcium sulfate). Order additional desiccant separately. Inlet and outlet ports accept 1/4 to 3/8 in. ID tubing Absorbs up to 50g of moisture
Specifications
Material Calcium Sulfate with Cobalt Chloride
Quantity 1 Ea.
Moisture 50g
Water Absorption 50g