missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ OTAL™ Hand Pump

Product Code. 10761002 Shop All Bürkle Products
Change view
Click to view available options
Flow Rate:
12 L/min.
14 L/min.
20 L/min.
26 L/min.
4 L/min.
Material:
PVDF
Polypropylene
Polypropylene, PVC
Stainless Steel
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Material Unit Size
10761002 20 L/min. Polypropylene Each
10697331 4 L/min. Polypropylene Each
10433291 12 L/min. Polypropylene, PVC Each
10652921 14 L/min. Polypropylene Each
10732492 20 L/min. PVDF Each
10270921 26 L/min. Polypropylene Each
10712332 26 L/min. Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Attention! Make sure to pay attention to the List of Chemical Resistance!
Product Code. 10761002 Supplier Buerkle™ Supplier No. 50051500
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Attention! Make sure to pay attention to the List of Chemical Resistance!

Ensure removal of residual liquid with these versatile hand pumps. Buerkle™ OTAL™ Hand Pumps feature a telescopic immersion tube that adapts itself to the container height on insertion, allowing the compact, space-saving hand pump to be used with different containers.

OTAL hand pumps fit most commercial storage drums. The telescopic immersion tube adapts itself to the container height on insertion. This ensures good removal of residual liquid and means that the hand pump can be used with different containers.
  • Particularly compact, space-saving hand pump
  • Good pump capacity
  • With universal bungs made of soft PVC, for container openings of 30–70 mm (1.18-2.76 in.) Ø
OTAL hand pumps with PP delivery tubes:
  • resistant to weak acids, alkalis and detergents.
Pump with PVDF delivery tubes:
  • particularly suitable for acids and alkalis, not for heavily oxidizing acids and alkalis.
  • With its special material properties, PVDF has an even better chemical resistance than PP. Resistant to chlorine bleach.
Stainless steel AISI 304 (1.4301) pump:
  • for liquids that are not easily combustible in Hazard Category AIII (VbF).
  • The OTAL PP or PVDF hand pump should be used for caustic media.
Pump with hose and stopcock:
  • discharge hose with a length of 1.5 m made of PVC and PP/PE stopcock, delivery tube made of PP.
  • The discharge hose is particularly suited to fill distant containers.
TRUSTED_SUSTAINABILITY

Specifications

Applications Commercial Storage
Flow Rate 20 L/min.
Includes 15 mm Dia. Polypropylene Delivery Tube
Weight (Metric) 25 kg
Depth (Metric) 70 cm
Pump Tube Length 70 cm
Resistance Properties Alkalis, Detergent, Weak Acid
Drum Opening Diameter 30 to 70 mm
Material Polypropylene
Product Type Hand Pump
For Use With (Equipment) Storage Drum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.