missing translation for 'onlineSavingsMsg'
Learn More

Tork Hand Sanitising Alcohol Foam S4

Product Code. 30243802 Shop All Tork Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30243802 Clear Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30243802 Supplier Tork Supplier No. 520102
  • ONLINE EXCLUSIVE PRICE
    163.00 €
    160.89 € / Pack of 6
    Save 2.11 €
    1% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dermatologically-tested, Tork hand sanitising alcohol foam is gentle on the skin.

  • Effective formula - EN 1500 and EN 14476.
  • This dermatologically tested foam is easy to handle and spreads easily
  • Save staff time with effortless refilling.
  • Help secure good hand hygiene - the factory-sealed bottle with a new pump for every refill helps reduce the risk of contamination and protect the formulations all the way to the hands.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Color Clear
Form Foam
Active Ingredients Alcohol
Packaging Plastic Bottle
Quantity 6/Pk.
Size 950 mL
compliance-icons compliance-icons
Product Identifier
  • Tork Hand Sanitizing Alcohol Foam
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.