Learn More
Gardner Denver Welch™ Handvac Professional Handpipettor Set
Product Code. 15351461 Shop All Welch Ilmvac Products
Change view
Click to view available options
For Use With (Application):
Aspiration
Unit Size:
Each
Product Code. For Use With (Application) Unit Size
15351461 Aspiration Each
1 options
Product Code. 15351461 Supplier Gardner Denver Welch™ Supplier No. 112580
  • ONLINE EXCLUSIVE PRICE
    528.00 €
    516.30 € / Pack of 1
    Save 11.70 €
    2% Off
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ergonomic and flexible handheld pipettor can be used in combination with pasteur pipettes, plastic aspiration pipettes and variety of other adapters.
Features:
  • Optimized vacuum level controlling
  • Suction regulation through press key on top of the HandVac
  • Comfortable vacuum hose connection
  • Perfect adaptation to human hand shape
  • Considers natural hand form with ergonomic features
  • Easy change of adapters and accessory
  • High quality components for trouble-free operation
  • Handvac is fully autoclavable
Includes:
  • Handvac handpipettor complete
  • 1-Channel Adapter for disposable tips
  • 8-Channel Adapter for disposable tips
  • 8-channel Stainless Steel adapter
  • Rubber adapter for Pasteur pipettes
  • 2m vacuum hose
  • Stand for handvac
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Aspiration
Product Line Handvac Professional Handpipettor Set
Type Complete Tool Set