missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Clear Tips, Presterilized
GREENER_CHOICE

Clear tips for accurate sample delivery

186.00 € - 345.00 €

Specifications

Sterility Sterile
Color Clear
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
For Use With (Application) For accurate sample delivery
View More Specs

Products 7
Product Code Brand Volume (Metric) Filtered Price Quantity & Availability  
Product Code Brand Volume (Metric) Filtered Price Quantity & Availability  
11354285
GREENER_CHOICE
View Documents Promotion Details
Axygen™
TT-1000-C-HTR-S
1000 μL Non-filtered
186.00 €
Pack of 1536
In Stock
to see stock.
Add to basket
 
11354295
GREENER_CHOICE
View Documents Promotion Details
Axygen™
TT-200-C-HTR-S
200 μL Non-filtered
278.00 €
Pack of 2304
Estimated Shipment: 27-03-2025
to see stock.
Add to basket
 
11304305
View Documents Promotion Details
Axygen™
TT-50-C-HTR-S
50 μL Non-filtered This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11344305
GREENER_CHOICE
View Documents Promotion Details
Axygen™
TTF-1000-C-HTR-S
1000 μL Filtered
230.00 €
Pack of 1536
Estimated Shipment: 21-11-2024
to see stock.
Add to basket
 
11384305
GREENER_CHOICE
View Documents Promotion Details
Axygen™
TTF-200-C-HTR-S
200 μL Filtered
345.00 €
Pack of 2304
In Stock
to see stock.
Add to basket
 
11364305
GREENER_CHOICE
View Documents Promotion Details
Axygen™
TTF-20-C-HTR-S
20 μL Filtered
345.00 €
Pack of 2304
Estimated Shipment: 27-03-2025
to see stock.
Add to basket
 
11304315
GREENER_CHOICE
View Documents Promotion Details
Axygen™
TTF-50-C-HTR-S
50 μL Filtered
329.00 €
Pack of 2304
Estimated Shipment: 27-03-2025
to see stock.
Add to basket
 
Description

Description

Axygen™ offers a comprehensive line of tips for Tecan™, Aurora Biomed and PerkinElmer™ automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are RNase-/DNase-free and nonpyrogenic. Available in nonsterile, sterile, non-filtered and filtered. Clear and Conductive black polypropylene tips are available in 96-well format and is also available in hanging tip rack format designed for Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head and PerkinElmer™ Janus™/MultiProbe with Variospan-1, 4, 8 head.


  • 1000 μL Clear Tips
  • 96 Tip format
  • Clear Polypropylene
  • RNase-/DNase-free
  • Nonpyrogenic
  • Sterile
  • Compatibility: Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head
  • Certifications

    Certified DNAse-, RNAse- and Pyrogen-safe

    Specifications

    Specifications

    Sterile
    Autoclavable
    For accurate sample delivery
    Automation, Round
    Clear
    Non-ANSI
    Rack
    Tecan Genesis, Tecan Freedom, Tecan Freedom EVO, Tecan Miniprep LiHa-1, 4, 8 Head, Aurora Biomed Versa 8-channel Head
    Videos
    SDS
    Documents

    Documents

    Product Certifications