missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Nonsterile
Black tips for accurate sample delivery
Brand: Axygen™ TT-50-CBK-HTR
Code : 66
Additional Details : Weight : 2.70000kg
Description
- Polypropylene
- Volume ranging from 20 to 1000μL
- Available with or without aerosol-resistant filters
- Compatible with Axygen™ Tecan™/Perkin Elmer/Auroroa™ Biomed/Corbett Cas-1200 systems
Specifications
50 μL | |
Non-sterile | |
Autoclavable | |
For accurate sample delivery | |
Automation, Round | |
Tecan Genesis, Tecan Freedom, Tecan Freedom EVO, Tecan Miniprep LiHa-1, 4, 8 Head, Aurora Biomed Versa 8-channel Head |
Non-Filtered | |
Black | |
Non-ANSI | |
Rack | |
Hanging Tip Racked | |
Polypropylene |