missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Nonsterile
Black tips for accurate sample delivery
Marca: Axygen™ TT-1000-CBK-HTR-24
Code : 66
Dettagli aggiuntivi : Weight : 3.18000kg
Descrizione
- Polypropylene
- Volume ranging from 20 to 1000μL
- Available with or without aerosol-resistant filters
- Compatible with Axygen™ Tecan™/Perkin Elmer/Auroroa™ Biomed/Corbett Cas-1200 systems
Specifica
1000 μL | |
Black | |
Non-ANSI | |
Rack | |
Hanging Tip Racked, Cardboard pack | |
Polypropylene |
Non-sterile | |
Autoclavable | |
For accurate sample delivery | |
Automation, Round | |
Tecan Genesis, Tecan Freedom, Tecan Freedom EVO, Tecan Miniprep LiHa-1, 4, 8 Head, Aurora Biomed Versa 8-channel Head |