Learn More
Corning™ Hank's Balanced Salt Solutions
Salt solutions are shipped at ambient temperature unless otherwise indicated
112.33 € - 125.00 €
Product Code | Brand | Concentration | Quantity | With Additives | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | With Additives | Price | Quantity & Availability | |||
15353571
|
Corning™
21-020-CV |
1 X | 1 x 500 mL | Calcium, Magnesium |
112.33 €
Pack of 6 |
Estimated Shipment: 25-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15373571
|
Corning™
21-022-CV |
1 X | 6 x 500 mL |
125.00 €
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15363571
|
Corning™
21-021-CV |
1 X | 6 x 500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
pH of 10X solutions must be adjusted following dilution to 1X concentration.
Specifications
Hanks' Balanced Salt Solution |