missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Checker™ pH Tester with 0.1 pH Resolution

Product Code. 10131361
Change view
Click to view available options
For Use With (Application):
pH
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
10131361 pH Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10131361 Supplier Hanna Instruments™ Supplier No. HI98103
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with virtually any electrode that shares same connector.

  • pH range from 0.00 to 14.00
  • Provides high accuracy with 0.1 pH resolution
  • Battery type 1.5V approximately 1000 hours of continous use
  • RH max 95%
  • With LCD and manual two point calibration
  • HI1271 pH electrode with screw type connector (included)

Specifications

For Use With (Application) pH
Accuracy ±0.2pH
Detection Range 0 to 14 pH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.