missing translation for 'onlineSavingsMsg'
Learn More

Hansa Yellow (Technical Grade), TRC

Product Code. 30179630
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179630 5 g 5g
30179603 1 g 1g
30179053 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179630 Supplier Toronto Research Chemicals Supplier No. TRCH1058185G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2512-29-0
Molecular Formula C17 H16 N4 O4
Molecular Weight (g/mol) 340.3333
Synonym Butanamide, 2-[(4-methyl-2-nitrophenyl)azo]-3-oxo-N-phenyl- (9CI), C.I. Pigment Yellow 1 (7CI,8CI), Hansa Yellow (6CI), 2-[2-(4-Methyl-2-nitrophenyl)diazenyl]-3-oxo-N-phenylbutanamide, ADC Pigment Yellow G, Accosperse Hansa Yellow G, Arylamide Yellow G, Arylide Yellow G, Brazil Yellow X 2866, Burma Yellow X 1622, C.I. 11680, Calcotone Hansa Yellow, Carnelio Yellow G, Chromatex Yellow J, Dainichi Fast Yellow G, Direct Yellow 161, Eljon Fast Yellow GN-GX, Eljon Fast Yellow PV Extra, Ext D and C Yellow No. 5, Fanchon Yellow G-YH 1, Fanchon Yellow WD 259, Fast Yellow 2070, Fast Yellow J, Fast Yellow JT, Fastona Yellow G, Fastona Yellow G transparent, Graphtol Yellow 4813-0, Graphtol Yellow GL, H 0312, Hancock Yellow 1008, Hansa Yellow G, Hansa Yellow G 02, Hansa Yellow G 41, Hansa Yellow G 45-4050, Hansa Yellow G Toner, Hansa Yellow G extra, Hansa Yellow GAD, Hansa Yellow GT, Hansa Yellow S 3155, Hansa Yellow Toner G-YA 8365, Helio Fast Yellow GN, Helio Fast Yellow GNS, Helio Fast Yellow GT, Heuco Yellow 100100, Irgalite Fast Yellow PG, Irgalite Fast Yellow PG transparent, Irgalite Yellow G, Irgalite Yellow GNX, Irgalite Yellow GTN, Irgalite Yellow PDS 8, Irgalite Yellow PV 2, Isol Aryl Yellow G, Isol Aryl Yellow G Transp., Isol Aryl Yellow GX, Isol Fast Yellow G, Isol Fast Yellow G Transp., Japan Yellow 401, Japan Yellow No. 401, Kromon Yellow G, Light yellow, Lightfast Yellow, Lightfast Yellow G, Lithosol Fast Yellow HL, Lithosol Fast Yellow HV, Lutetia Fast Yellow J, Lutetia Yellow J 711, Makicolor WD Yellow, Mono Fast Yellow G, Monolite Fast Paper Yellow GN, Monolite Fast Yellow 2RA, Monolite Fast Yellow GA, Monolite Fast Yellow GN, Monolite Fast Yellow GNA, Monolite Fast Yellow GNV, Monolite Yellow GN, NSC 6131, No. 9 Forthfast Yellow G, No. 9/10 Forthfast Yellow G, No. 9/4604 Forthfast Yellow G, Oralith Yellow G, Oriental Yellow GT, Permanent Yellow Dark YT 392, Permanent Yellow G, Permanent Yellow G Toner, Permansa Yellow G, Pigment C.I. 11680, Pigment Fast Yellow, Pigment Lightfast Yellow, Pigment Lightfast Yellow TP, Pigment Lightfast Yellow VCO, Pigment Yellow 1, Pigment Yellow G, Pigment Yellow Lightfast VCO, Recolite Fast Yellow G, Sanyo Fast Yellow G, Segnale Light Yellow AG, Segnale Light Yellow AGC, Segnale Light Yellow G, Segnale Light Yellow GM, Sico Yellow D 1250, Sico Yellow L 1250, Siegle Yellow G, Siegle Yellow GTR transparent, Silopol Yellow G, Silosol Yellow GN, Siloton Yellow GCL, Symuler Fast Yellow GH, Syton Fast Yellow G, Talens Yellow, Tertropigment Yellow G, Toluidine Yellow, Toluidine Yellow AH 175, Toluidine Yellow G, Transa Flo Yellow, Versal Yellow G, Vulcafix Yellow 4J, Yellow Fast 23, Yellow Pigment G, α-(o-Nitro-p-tolylazo)acetoacetanilide
IUPAC Name 2-[(E)-(4-methyl-2-nitrophenyl)diazenyl]-3-oxo-N-phenylbutanamide
SMILES CC(=O)C(N=Nc1ccc(C)cc1[N+](=O)[O-])C(=O)Nc2ccccc2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C17H16N4O4/c1-11-8-9-14(15(10-11)21(24)25)19-20-16(12(2)22)17(23)18-13-6-4-3-5-7-13/h3-10,16H,1-2H3,(H,18,23)
Formula Weight 340.1172
Chemical Name or Material Pigment Yellow 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.