missing translation for 'onlineSavingsMsg'
Learn More

3M™ Hard Hat Integrated Safety Glasses V9C

Product Code. 17237640
missing translation for 'orderingAttributeHoverText'
:
20 /Cs.
This item is not returnable. View return policy

Product Code. 17237640

missing translation for 'mfr': 3M™ 7100092589

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Clear lens safety glasses designed to attach to 3M™ Hard Hats for protection from particles and UV.

  • Provides eye protection from particles and UV
  • Easily attaches to 3M™ Hard Hats
  • Durable anti-scratch and anti-fog lens coating helps minimise physical damage and fogging
  • Easy to adjust for a good fit
  • Tested and approved to EN 166:2001
  • Faceshield marking 2C-1,2 3M 1 F
TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Lens Tint Clear
For Use With (Equipment) 3 M™ Hard Hats
For Use With (Application) Protection From Particles and UV
Quantity 20 /Cs.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.