Learn More
Brady™ Harsh Environment Multipurpose Continuous Opaque Polyester Labels for M6/M7 Printers
Permanent polyester is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
Brand:  Brady™ 173228
 View more versions of this product
Product Code. 17898994
  • ONLINE EXCLUSIVE PRICE
    50.80 €
    49.24 € / Pack of 1
    Save 1.56 €
    3% Off
In Stock
Add to basket
Description
Description
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • UL approved, CSA accepted
  • Meets requirements of a halogen-free material per DIN VDE 0472 part 815
  • Service temperature of -70°C to 110°C (-94°F to 230°F)
  • Made of B-423 polyester material
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
White
Continuous
Single-side Printable
Label
Barcode Labeling, Circuit Board Labeling, Equipment Labeling, General Identification, Inventory and Inspection Labeling
No
Rectangle
-70°C to +110°C
0.076 mm
50 ft.
0.5 in.
M7C-500-423
Halogen-free (DIN VDE 0472 Part 815)
None
Gloss
Polyester
Thermal Transfer
Non-sterile
M710, BMP71
50 ft.
-94°F to +230°F
0.003 in.
50 ft. x 0.5 in.
15.24 m
12.7 mm