Learn More
Brady™ Harsh Environment Multipurpose Die-cut Clear Polyester Labels for M6/M7 Printers
Get aggressive adhesive labels for difficult surfaces, and the harshest environments. These clear, polyester labels are ideal for labs, asset tracking and faceplate labeling.
Brand:  Brady™ 174098
 View more versions of this product
Product Code. 17898444
  • ONLINE EXCLUSIVE PRICE
    83.70 €
    81.27 € / Pack of 100
    Save 2.43 €
    2% Off
Estimated Shipment: 06-11-2026
Add to basket
Description
Description
  • High-tack permanent adhesive for stainless steel, powder coated metals, painted enamel, and plastics in all-weather applications
  • Resists exposure to UV light, extreme temperatures, fuel and oil, degreasers, cleaners, and detergents
  • Black print on clear polyester
  • UL Recognized to UL969, CSA to C222 No 015-95, RoHS compliant to 2011/65/EU (Pass 2015/863)
Specifications
Specifications
Autoclavable
Permanent Acrylic
Clear
Die-cut
Single-side Printable
Label
Asset Tracking, Barcode Labeling, Equipment Labeling, Electrical Labeling, General Identification
No
Rectangle
-70°C to +100°C
0.076 mm
3 in.
25.4 mm
M6-22-430
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, CSA Approved, UL Recognized
None
Gloss
Polyester
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
100 Labels
-94°F to +212°F
0.003 in.
3 x 1 in.
1 in.
76.2 mm