Learn More
Thermo Scientific™ Harvest RC, BC1020
Product Code. 31767521 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Chromatography Filter
Unit Size:
Each
Product Code. Type Unit Size
31767521 Chromatography Filter Each
1 options
Product Code. 31767521 Supplier Thermo Scientific™ Supplier No. 7100236866
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Harvest RC Chromatographic Clarifier is a single stage single-use chromatographic clarification solution designed for harvesting and clarification of modern cell cultures. Clarification with this solution offers fewer processing steps, higher yield and lower production time and costs.
  • Simplification of high cell density cell culture fluid clarification unit operations.
  • Remove cells, cell debris and DNA
  • Optimized for PCV high density CHO cell culture (5-8% PCV)
  • Replacement of primary, secondary and guard membrane clarification stages.
  • Typical product recovery of 95+% (capsules)
  • Synthetic chromatographic harvest media with chemically defined extractables.
  • Predictable scaling from discovery to manufacturing.
  • Lower total cost of manufacturing compared to centrifugation and depth filtration.
  • Lower consumption of buffer and water compared to depth filtration.
  • Capsules fit into laboratory to manufacturing scale workflows.
Specifications
Disposable Single-use
Filter Type Filter Capsule
Material (Membrane) PES
Sterility Non-sterile
Quantity 1 Ea.
Type Chromatography Filter