Learn More
3M™ Harvest RC, BC25, Luer
Product Code. 31767513
Change view
Click to view available options
Type:
Chromatography Filter
Unit Size:
Pack of 2
Product Code. Type unitSize
31767513 Chromatography Filter Pack of 2
1 options
Product Code. 31767513 Supplier 3M™ Supplier No. EMP301HRC2FA
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
The 3M™ Harvest RC Chromatographic Clarifier is a single stage single-use chromatographic clarification solution designed for harvesting and clarification of modern cell cultures. Clarification with this solution offers fewer processing steps, higher yield and lower production time and costs.
  • Simplification of high cell density cell culture fluid clarification unit operations.
  • Remove cells, cell debris and DNA
  • Optimized for PCV high density CHO cell culture (5-8% PCV)
  • Replacement of primary, secondary and guard membrane clarification stages.
  • Typical product recovery of 95+% (capsules)
  • Synthetic chromatographic harvest media with chemically defined extractables.
  • Predictable scaling from discovery to manufacturing.
  • Lower total cost of manufacturing compared to centrifugation and depth filtration.
  • Lower consumption of buffer and water compared to depth filtration.
  • Capsules fit into laboratory to manufacturing scale workflows.
Specifications
Disposable Single-use
Filter Type Filter Capsule
Material (Membrane) PES
Sterility Non-sterile
Quantity 2/Cs.
Type Chromatography Filter