Learn More
Thermo Scientific™ Harvest RC Centrate Chromatographic Clarifier, BC340
Shop All Thermo Scientific ProductsDescription
- Single-stage centrate clarification for process compression and streamlining of process development.
- Enables de-risking of clarification process development, scale-up and tech transfer for faster speed to market.
- Synthetic, functionalized fibrous media provides effective soluble impurity removal in centrate, including DNA and cell shear-related submicron particle reduction.
- Predictable performance scalability for consistent centrate clarification at various process scales.
- The single-mode electrostatic separation mechanism and high-throughput media enable product recoveries of >95%.
- Provides comparable effluent quality to Harvest RC Chromatographic Clarifier for a modern clarification platform.
- Lower consumption of buffer or water compared to depth filtration.
- Alkaline-resistant capsules for pre-use sanitization.
Specifications
Specifications
| Disposable | Single-use |
| Filter Type | Filter Capsule |
| Material (Membrane) | Polyamide |
| Sterility | Non-sterile |
| Quantity | 1 Ea. |
| Type | Chromatography Filter |