Learn More
Thermo Scientific™ Harvest RC, CT15
Product Code. 31767517 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Chromatography Filter
Unit Size:
Pack of 12
Product Code. Type Unit Size
31767517 Chromatography Filter Pack of 12
1 options
Product Code. 31767517 Supplier Thermo Scientific™ Supplier No. 7100245350
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Harvest RC Chromatographic Clarifier is a single stage single-use chromatographic clarification solution designed for harvesting and clarification of modern cell cultures. Clarification with this solution offers fewer processing steps, higher yield and lower production time and costs.
  • Simplification of high cell density cell culture fluid clarification unit operations.
  • Remove cells, cell debris and DNA
  • Optimized for PCV high density CHO cell culture (5-8% PCV)
  • Replacement of primary, secondary and guard membrane clarification stages.
  • Typical product recovery of 95+% (capsules)
  • Synthetic chromatographic harvest media with chemically defined extractables.
  • Predictable scaling from discovery to manufacturing.
  • Lower total cost of manufacturing compared to centrifugation and depth filtration.
  • Lower consumption of buffer and water compared to depth filtration.
  • Capsules fit into laboratory to manufacturing scale workflows.
Specifications
Disposable Single-use
Filter Type Filter Capsule
Material (Membrane) PES
Sterility Non-sterile
Quantity 12/Cs.
Type Chromatography Filter
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.