missing translation for 'onlineSavingsMsg'
Learn More

Hastelloy C∣r gauze, 40 mesh woven from 0.19mm (0.0075in) dia wire

Product Code. 15447285
Change view
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15447285 300 x 300 mm Each
15427285 75 x 75 mm Each
15437285 150 x 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15447285 Supplier Thermo Scientific Alfa Aesar Supplier No. 046690.HB
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hastelloy C is used in combustion chambers, heat-treating equipment, collector rings, combustion chambers and exhaust stacks for high-heat applications in for conventional aircraft and jets, pipe connections, pumps and valves for oil refinery, chemical and petrochemical applications. Also used in components exposed to flue gas or flue gas desulfurization plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hastelloy C™ gauze
Name Note 40 mesh woven from 0.19mm (0.0075 in.) dia. wire
Quantity 300 x 300 mm
Mesh Size 40 Mesh
Form Wire Cloth
MDL Number MFCD00801065
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.