missing translation for 'onlineSavingsMsg'
Learn More

HATU, 97%

Product Code. 13438528 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13438528 25 g 25g
13219522 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13438528 Supplier Thermo Scientific Acros Supplier No. 365310250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 148893-10-1 | C10H15F6N6OP | 380.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148893-10-1
Molecular Formula C10H15F6N6OP
Molecular Weight (g/mol) 380.24
MDL Number MFCD00274639
InChI Key JNWBBCNCSMBKNE-UHFFFAOYSA-N
SMILES F[P-](F)(F)(F)(F)F.CN(C)C(ON1N=NC2=CC=CN=C12)=[N+](C)C

Specifications

Melting Point 189.0°C to 191.0°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 4653
IUPAC Name [bis(dimethylamino)methylidene]({3H-[1,2,3]triazolo[4,5-b]pyridin-3-yl})oxidanium
Formula Weight 380.23
Chemical Name or Material HATU
compliance-icons compliance-icons
Product Identifier
  • HATU
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.