missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Hazard Warning Tape
Click to view available options
Display Text:
Biohazard
Danger Broken Glass
Dimensions (LxW):
66 m x 25 mm
66 m x 50 mm
Unit Size:
Each
Description
Repeats warning with hazard symbol for up to 66m. Azpack™ Hazard Warning Tape is self-adhesive and printed black on a yellow background.
- Self-adhesive tape
- Wording and hazard symbol repeated along tape
- Black on yellow background
- 66m roll
- Cardboard core diameter: 75mm
- Width: 25mm
Specifications
Specifications
| Color | Black, Yellow |
| Format | Roll |
| Material | Vinyl |
| Display Text | Danger Broken Glass |
| Quantity | 1 Ea. |
| Dimensions (LxW) | 66 m x 50 mm |
| Length (Metric) | 66 m |
| Width (Metric) | 50 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction