missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Hazardous Spill Response Kit
Click to view available options
Capacity (Metric):
5 L
26 L
Dimensions (L x W):
400 x 70 mm
500 x 630 mm
Includes:
10 sheets, 1 pillow, 1 disposable bag with tie
10 sheets, 5 pillows, 5 mini-booms and 2 disposable bags with ties.
Unit Size:
Each
Description
Kits contain chemical sorbents made from inert, synthetic fibers to be deployed wherever hazardous liquids are in use, for the protection of both employees and equipment. In the event of a spill, the kit can be used to contain and clean up the hazardous liquid without delay.
- Constructed from polypropylene fibers
- Available in a wide range of formats
- Lightweight and dust-free
- High absorption capacity minimizes waste for disposal.
Specifications
Specifications
| Capacity (Metric) | 26 L |
| Material | Polypropylene Fiber |
| Type | Spill Kit |
| Includes | 10 sheets, 5 pillows, 5 mini-booms and 2 disposable bags with ties. |
| Packaging Quantity | 1 Ea. |
| Dimensions (L x W) | 500 x 630 mm |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion