missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Hazardous Substance Labels, 76 mm Core

Product Code. 15961021 Shop All Brady Products
Change view
Click to view available options
Includes:
1 Diamond Layout
2 Diamond Layout
3A Diamond Layout
3B Diamond Layout
4A Diamond Layout
4B Diamond Layout
Unit Size:
Pack of 1000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
15961021 3A Diamond Layout Pack of 1000
15941021 1 Diamond Layout Pack of 1000
15951021 2 Diamond Layout Pack of 1000
15971021 3B Diamond Layout Pack of 1000
15981021 4A Diamond Layout Pack of 1000
15991021 4B Diamond Layout Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15961021 Supplier Brady™ Supplier No. 622285
  • ONLINE EXCLUSIVE PRICE
    327.00 €
    313.38 € / Pack of 1000
    Save 13.62 €
    4% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pre-printed, benchtop printer labels for CLP-GHS hazardous substances.

  • Supplied with pre-printed red diamonds
  • Ready for you to print the required warning, compliant to the latest regulations

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color Red on White
Finish Gloss
Format Die-cut
Material Polypropylene (B-7594)
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Hazard Communication Identification
For Use With (Equipment) i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Includes 3A Diamond Layout
Labels per Row 1
Min. Operating Temperature (Metric) -20°C
Model THTCLP-M-L3A-7594-1
No. per Pack 1000 Label(s) / Roll
Quantity 1000 Labels
Shape Rectangle
Size 75.00 mm (W) x 110.00 mm (H)
Temperature Range (English) -4°F to 212°F
Temperature Range (Metric) -20°C to +100°C
Thickness (English) 0.0031 in.
Thickness (Metric) 0.079 mm
Vertical Repeat (Metric) 115.4 mm
Dimensions (L x W) 75.00 mm (W) x 110.00 mm (H)
Length (English) 4.33 in.
Width (English) 2.95 in.
Width (Metric) 75 mm
Width (English) Printable Area 75.00 mm
Height (English) 4.33 in.
Height (Metric) 110 mm
Alternate Part Number THTCLP-M-L3A-7594-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.