missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Hazardous Substances Rack

Product Code. 17866556 Shop All asecos Products
Change view
Click to view available options
Capacity (Metric):
25 L
40 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17866556 25 L Each
17824625 40 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17866556 Supplier asecos™ Supplier No. 27833
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Hazardous Substances Rack provides storage of water pollutants with a steel sump on each storage level.

  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars
  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Rack system can be extended individually
  • Flat-pack delivery with assembly instructions
  • 4 painted sumps made of steel as storage levels

Specifications

Capacity (English) 6.6 gal.
Capacity (Metric) 25 L
Material Steel
Type Drum Rack
Length (English) 53.54 in.
Length (Metric) 1360.00 mm
Width (English) 53.54 in.
Width (Metric) 1360.00 mm
Height (English) 78.74 in.
Height (Metric) 2000.00 mm
Depth (English) 17.20 in.
Depth (Metric) 437.00 mm
Weight (English) 182.98 lb.
Weight (Metric) 83 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.