missing translation for 'onlineSavingsMsg'
Learn More

Woodsafe™ Sharpsbin and Hazardous waste containers with yellow single lid

Product Code. 18176539
Change view
Click to view available options
Capacity (Metric):
6 L
12 L
50 L
Unit Size:
Each
Pack of 20
Pack of 30
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
18176539 50 L Each
18742825 6 L Pack of 30
18702825 12 L Pack of 20
18136539 - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Follow instructions for safe and correct use
Product Code. 18176539 Supplier Woodsafe™ Supplier No. 1500212
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Follow instructions for safe and correct use

Woodsafe is a cost-effective solution for reducing the use of fossil based plastic by 80% and CO2 emissions by 66%, documented in in a full LCA. Produced with residue from the forest industry using green energy. Available in sizes from 0.5 L to 50 L. Single and double lid versions.

  • Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the Swedish forest industry
  • Lowers the use of Fossil-based plastic by 80%!
  • Lowers the carbon footprint by more than 60%, proved in full LCA
  • Made from 100% Biocomposite
  • The lid can be securely permanently closed for transportation to the final disposal
  • My Green Labs ACT Label
  • Follows same process as yellow bins and can be used to transport the same substances
  • Natural color and smell from wood - No migration of substances
  • Compliant with UN3291
  • Tested according to ISO 23907-1:2019
  • UN ADR Approved
  • Biocomposite have the same properties as fossil-based plastic and can be used exactly the same way
  • NOTE: String between lids is for delivery purpose, remove/break before use!
  • Stop burning plastic

Specifications

Capacity (Metric) 50 L
Certifications/Compliance EN ISO 23907-1:2019. UN3291, ADR Approved
Closure Type Permanent closure
Closure Color Yellow
Capacity (English) 11.87 gal
Color Brown, Yellow
Dimensions (L x W x H) 15.6 x 15.6 x 19 in.
Disposable Single-use
Stackable Yes
Sterility Non-sterile
Autoclavable Autoclavable
Includes Bin, Lid, and Label
For Use With (Application) Safe disposal of sharps and potentially infectious waste or medical waste
Packaging Cardboard
Material Biocomposite
Product Type Biohazard Waste Container
Show More Show Less

Lower climate impact and ACT label

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.