Learn More
Zeiss™ HBO 100 Illuminator
Product Code. 11835440
Change view
Click to view available options
Type:
HBO 100 Illuminator
Unit Size:
Each
Product Code. Type Unit Size
11835440 HBO 100 Illuminator Each
1 options
Product Code. 11835440 Supplier Zeiss™ Supplier No. 4230119901000
  • ONLINE EXCLUSIVE PRICE
    4540.00 €
    3883.34 € / Pack of 1
    Save 656.66 €
    14% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Features automatic lamp alignment
  • HBO 100 illuminator
  • With collector and lamp mount
  • Includes heat-reflecting filter
  • Automatic Lamp Alignment
Specifications
Type HBO 100 Illuminator
Description Features automatic lamp alignment
For Use With (Equipment) Zeiss Microscopes
Includes Lamp Mount and Collector