missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HBSS (10X), calcium, magnesium, no phenol red
GREENER_CHOICE

Product Code. 11530466
Click to view available options
Quantity:
500 mL
10 x 500 mL
Unit Size:
5000mL
500mL
This item is not returnable. View return policy

Product Code. 11530466

Brand: Gibco™ 14065049

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

HBSS (10X), calcium, magnesium, no phenol red

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ HBSS product made in our Grand Island facility (14065-056). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Concentrated forms of Gibco™ HBSS require pH adjustment and filtration at the time of preparation.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 13485 standards, FDA
Content And Storage 15 to 30°C
Description Hanks' Balanced Salt Solution, 500mL, Liquid, 10X Concentration, 5.5 to 6.0 pH, With Calcium, Magnesium, Glucose, 36 Months Shelf Life, For Mammalian Cell Culture
Concentration 10 X
Form Liquid
Product Type Hanks' Balanced Salt Solution
With Additives Calcium, Magnesium, Glucose
Without Additives No Sodium Bicarbonate, No Phenol Red
For Use With (Application) Mammalian Cell Culture
pH 5.5 to 6.0
Quantity 500 mL
Regulatory Status For in-vitro diagnostic use
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.