missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HBSS, calcium, magnesium, phenol red

Product Code. 30276209
Change view
Click to view available options
Quantity:
5 L
20 L
50 L
100 L
200 L
Format:
Drum
Sample
Tote
Unit Size:
100 L
20 L
200 L
50 L
Each
Product Code. Quantity Format unitSize
30276209 5 L Sample Each
30276692 20 L Tote 20 L
30275726 50 L Drum 50 L
30276439 100 L Drum 100 L
30276690 200 L Drum 200 L
5 options
This item is not returnable. View return policy

Product Code. 30276209

Brand: Gibco™ A59772BA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco™ Balanced Salt Solutions are sterile-filtered, packaged in bioprocess containers, and ready to order.

Gibco™ Balanced Salt Solutions are sterile-filtered, packaged in bioprocess containers, and ready to order. Commonly used for processing cells and flushing equipment, sourcing Gibco balanced salt solutions can help streamline your operation by reducing buffer preparation and allowing you to focus on additional critical steps in your biopharmaceutical processes. Discover new outsourcing efficiencies to reclaim time, space, and resources.

Product Features

  • Multiple formulations and concentrations available
  • Available in a range of pack sizes (5 L, 20 L, 50 L, 100 L, and 200 L)
  • Established products with set specifications are made to order
  • Product standardization enables shorter times from inquiry to quote and shipment, compared to custom
  • Readily accessible documentation for accelerated qualification

Benefits

  • Reduces time spent preparing buffers in-house
  • Mitigates space requirement for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Meets workflow needs across stages of development and manufacturing of biologicals
TRUSTED_SUSTAINABILITY

Specifications

Color Clear, colorless solution
Form Liquid
Product Type Hanks' Balanced Salt Solution (HBSS)
Sterility Sterile-filtered
Without Additives No Calcium, No Magnesium
Format Sample
Manufacturing Quality cGMP
Osmolality 265 to 295 mOsm/kg
pH 6.7 to 7.8
Quantity 5 L

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.