missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HBSS, no calcium, no magnesium
GREENER_CHOICE

Product Code. 11500476
Change view
Click to view available options
Quantity:
100 mL
500 mL
10 x 500 mL
20 x 100 mL
Unit Size:
1 set
100mL
500mL
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11500476 10 x 500 mL 1 set
11580466 100 mL 100mL
11590466 500 mL 500mL
11510476 20 x 100 mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11500476 Supplier Gibco™ Supplier No. 14170138

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

HBSS, no calcium, no magnesium

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ HBSS product made in our Grand Island facility (14170-120). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 13485 standards, FDA
Content And Storage 15 to 30°C
Description Hanks' Balanced Salt Solution, 10 x 500mL, Liquid, 1 X Concentration, 7.0 to 7.4 pH, With Glucose, Sodium Bicarbonate, Phenol Red, 36 Months Shelf Life, For Mammalian Cell Culture
Concentration 1 X
Form Liquid
Product Type Hanks' Balanced Salt Solution
With Additives Glucose, Sodium Bicarbonate, Phenol Red
Without Additives No Calcium, No Magnesium
For Use With (Application) Mammalian Cell Culture
pH 7.0 to 7.4
Quantity 10 x 500 mL
Regulatory Status For in-vitro diagnostic use
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.