missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ HBX 41108

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Inhibitor of ubiquitin-specific protease (USP) 7 activity (IC50 = 424 nM). Displays uncompetitive inhibition. Also inhibits USP7-mediated p53 deubiquitination (IC50 = 0.8 μM). Stabilizes p53 and inhibits cancer cell growth; induces p53-dependent apoptosis in p53 wild type and null isogenic cancer cell lines.
Specifications
Specifications
| CAS | 924296-39-9 |
| Quantity | 10 mg |
| Molecular Formula | C13H3ClN4O |
| Formula Weight | 266.64 |
| Purity | >98% |
| Molecular Weight (g/mol) | 266.64 |
| Chemical Name or Material | HBX 41108 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction