Learn More
Invitrogen™ HCS CellMask™ Stains
Description
Identify entire cells (cytoplasm and nucleus) during high-content screening (HCS) assays with the versatile HCS CellMask Red, CellMask Orange, CellMask Green, CellMask Blue, CellMask Deep Red, and CellMask Near-IR stains, which are fluorophores that can be used immediately after fixation and permeabilization, or after antibody labeling. They are available in a variety of colors for multiplexing protocols.
HCS CellMask stains can be applied to cells immediately after fixation or in the last step of multiplexing protocols, and they are compatible with detergent-based cell permeabilization protocols. If only nuclear staining is desired, HCS NuclearMask stains can be used for measuring DNA content in addition to enabling robust cell demarcation. HCS NuclearMask stains are also available in a choice of colors for multiplexing flexibility.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Color | Far-Red, Deep Red |
| Content And Storage | Includes 1 set of vials, providing sufficient material to stain a total of ten 96-well microplates. Store at ≤-20°C, desiccate, and protect from light. |
| Excitation Wavelength Range | 650/655 |
| For Use With (Equipment) | High Content Instrument |
| Quantity | 1 Set |
| Detection Method | Fluorescence |
| Description | HCS CellMask Deep Red Stain |
| Emission | Near IR |
| Product Line | CellMask |
| Shipping Condition | Room Temperature |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.