missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals HCV Blot 3.0 (CE)

Product Code. 30694942 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
18 Tests/Kit
36 Tests/Kit
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30694942 36 Tests/Kit 1 set
30694927 18 Tests/Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30694942 Supplier MP Biomedicals Supplier No. 11130036
  • ONLINE EXCLUSIVE PRICE
    707.42 €
    673.73 € / 1 sets
    Save 33.69 €
    4% Off
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The Blot is a qualitative enzyme immunoassay for the in vitro detection of antibodies to hepatitis C virus (HCV) in human serum or plasma.

  • The nitrocellulose strips are coated with four recombinant HCV proteins from the Capsid, NS3, NS4, and NS5 regions of the HCV genome.
  • The HCV proteins are expressed as GST fusion proteins, so a GST control band is included to indicate reactivity to native GST.
  • The blots also contain an IgG control band and an anti-IgG band. Individual nitrocellulose strips are incubated with diluted serum or plasma specimens and controls.
  • Specific antibodies to HCV, if present in the specimen, will bind to the HCV proteins on the strips.
  • The strips are washed to remove unbound materials and then incubated with affinity purified anti-human IgG conjugated with alkaline phosphatase.
  • The conjugate antibody will bind to any antigen-antibody complexes formed on the blots.
  • Unbound conjugate is removed by washing. A BCIP/NBT substrate is added to visualise reactive protein bands on the blots.
  • The HCV BLOT 3.0 includes structural and non-structural antigens from HCV and is intended as supplemental confirmatory assay for presence of antibodies to HCV.
  • It is intended for use as a more specific supplemental test on specimens found repeatedly reactive using specific procedures such as ELISA.
  • CE-marked.
  • For professional use only.
  • Human serum and plasma.

Specifications

Product Type Western Blot Kit
Detection Method Colorimetric
For Use With (Application) Immunoblot (Western)
Membrane Compatibility Nitrocellulose
Quantity 36 Tests/Kit
Substrate Alkaline Phosphatase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.