missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ HCV Proteome Microarray

Product Code. 16357758
Change view
Click to view available options
Quantity: 1 Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16357758 Quantity: 1 Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16357758 Supplier PEPperCHIP™ Supplier No. SPC.013.001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

HCV Proteome Microarray with 3084 virus specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.

  • Microarray Content: Genome polyprotein of hepatitis C virus cleaved into 11 chains: core protein p21, core protein p19, envelope glycoprotein E1, envelope glycoprotein E2 , p7, Protease NS2-3, serine protease NS3, non-structural protein 4A, Non-structural protein 4B, Non-structural protein 5A, RNA-directed RNA polymerase
  • Peptide Length/Overlap: 15 aa/ 12 aa
  • Number of Peptides: 3084 printed in duplicate and HA, polio and c-Myc control peptides

Specifications

Coating PEGMA/PMMA
Description 1 array copy per microarray
Quantity 1 Slide
Method of Attachment Combinatorial peptide synthesis by amino acid laser printing
Thickness (Metric) 1 mm
Certifications/Compliance ISO 9001:2015
Color Transparent
Regulatory Status RUO
Barcode Individual barcode for each microarray
Material Microscopic Glass
Type High-density Peptide Microarray
For Use With (Application) Antibody validation, epitope mapping, serum screening, biomarker discovery
Sterility Non-sterile
Dimensions (LxW) 75.4 x 25 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.