Learn More
SymLine Safety Waste Caps
Product Code. 16305988
Change view
Click to view available options
Size:
GL 45, GL 25
GL45
S60/61
Includes:
2 x PFA-fittings
4 x PFA-fittings
4 x PFA-fittings, 1 x tube connector
7 x PFA-fittings
Electronic fill level control
Filling Lance, Connector for exhaust filter
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Includes Unit Size
16305988 GL 45, GL 25 Filling Lance, Connector for exhaust filter Each
17172566 GL45 2 x PFA-fittings Each
17192566 GL45 4 x PFA-fittings Each
17102576 GL45 4 x PFA-fittings, 1 x tube connector Each
17182566 GL45 7 x PFA-fittings Each
10820411 S60/61 Electronic fill level control Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16305988 Supplier SymLine Supplier No. 106527
  • ONLINE EXCLUSIVE PRICE
    146.00 €
    133.97 € / Pack of 1
    Save 12.03 €
    8% Off
View all promotions
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Safety Waste Caps safely connect the pipework to the waste container. The integrated exhaust filter blocks solvent vapours – for the protection of health and the environment.
  • Safety Waste Caps are available with different thread sizes for attachment to the pipe system and with different numbers of hose connections for disposal of the liquid waste.
  • Optionally, Safety Waste Caps are available with electronic level sensors that indicate the degree of filling of the disposal container.
  • By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
  • Specifications
    For Use With (Equipment) SymLine System
    Includes Filling Lance, Connector for exhaust filter
    Material Polyphenylene Sulfide, PFA, PTFE
    Size GL 45, GL 25
    For Use With (Application) Waste Disposal System
    Product Type Safety Waste Cap