missing translation for 'onlineSavingsMsg'
Learn More

SCAT Collecting Tray, HDPE-EC

Product Code. 10530251 Shop All SCAT Europe Products
Click to view available options
Capacity (Metric):
10.5 L
12 L
23 L
24 L
Color:
Black
White
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10530251

Brand: SCAT 117983

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

SCAT Collecting Trays store your waste containers and provide additional safety in case of any leakage.

  • Suitable for a wide range of canisters
  • Collect liquid in case of a leakage
  • Available with base insert so the canisters stay always clean and dry
  • Optional with electrostatic conductive material
  • Made of PE-HD or optional PE-HD-ec

Specifications

Capacity (Metric) 12 L
Dimensions (L x W x H) Inner: 335 x 235 x 160 mm - Outer: 400 x 300 x 170 mm
Description Collecting tray, PE-HD, dimensions (W x H x D): 235 x 160 x 335 mm (interior), dimensions (W x H x D): 300 x 170 x 400 mm (exterior)
Color White
Height (Metric) 160 mm (Inner), 170 mm (Outer)
Length (Metric) 335 mm (Inner), 400 mm (Outer)
Material HDPE
Type Collection Tray
Width (Metric) 235 mm (Inner), 300 mm (Outer)
For Use With Waste Containers
For Use With (Application) Handling of Hazardous Substances
Quantity 1 Pc.
Shape Rectangular
Autoclavable Not Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.