missing translation for 'onlineSavingsMsg'
Learn More

SCAT Collecting Tray, HDPE-EC

Product Code. 10569290 Shop All SCAT Europe Products
Change view
Click to view available options
Capacity (Metric):
10.5 L
12 L
23 L
24 L
Color:
Black
White
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color unitSize
10569290 10.5 L Black Each
10334320 12 L Black Each
10530251 12 L White Each
10282710 23 L White Each
10222480 24 L Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10569290 Supplier SCAT Supplier No. 117985
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

SCAT Collecting Trays store your waste containers and provide additional safety in case of any leakage.

  • Suitable for a wide range of canisters
  • Collect liquid in case of a leakage
  • Available with base insert so the canisters stay always clean and dry
  • Optional with electrostatic conductive material
  • Made of PE-HD or optional PE-HD-ec

Specifications

Capacity (Metric) 10.5 L
Dimensions (L x W x H) Inner: 385 x 285 x 95 mm - Outer: 445 x 355 x 135 mm
Description Collecting tray with base insert, PE-HD electrostatic conductive, dimensions (W x H x D): 285 x 95 x 385 mm (interior), dimensions (W x H x D): 355 x 135 x 445 mm (exterior)
Color Black
Height (Metric) 135 mm (Outer), 95 mm (Interior)
Length (Metric) 445 mm (Outer), 385 mm (Inner)
Material HDPE
Type Collection Tray
Width (Metric) 285 (Inner) mm, 355 mm (Outer)
For Use With Waste Containers
For Use With (Application) Handling of Hazardous Substances
Includes Base Insert
Quantity 1 Pc.
Finish Electrically Conductive HDPE
Shape Rectangular
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.