missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT Collecting Tray, HDPE-EC
SCAT Collecting Trays store your waste containers and provide additional safety in case of any leakage.
95.85 € - 394.00 €
Specifications
Material | HDPE |
---|---|
Type | Collection Tray |
For Use With | Waste Containers |
For Use With (Application) | Handling of Hazardous Substances |
Shape | Rectangular |
Product Code | Brand | Capacity (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Color | Price | Quantity & Availability | ||||
10569290
|
SCAT
117985 |
10.5 L | Black |
354.00 €
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
10334320
|
SCAT
108981 |
12 L | Black |
262.00 €
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
10530251
|
SCAT
117983 |
12 L | White |
95.85 €
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
10282710
|
SCAT
117984 |
23 L | White |
111.00 €
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
10222480
|
SCAT
117986 |
24 L | Black |
394.00 €
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
Description
- Suitable for a wide range of canisters
- Collect liquid in case of a leakage
- Available with base insert so the canisters stay always clean and dry
- Optional with electrostatic conductive material
- Made of PE-HD or optional PE-HD-ec
Specifications
HDPE | |
Waste Containers | |
Rectangular |
Collection Tray | |
Handling of Hazardous Substances | |
Not Autoclavable |