missing translation for 'onlineSavingsMsg'
Learn More

SymLine™ Thread Adapter, Type 14

Product Code. 15342352
Change view
Click to view available options
Closure Size:
G 1 1/2 in., 32 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Unit Size
15342352 G 1 1/2 in., 32 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15342352 Supplier SymLine™ Supplier No. 106434
  • ONLINE EXCLUSIVE PRICE
    78.25 €
    68.81 € / Pack of 1
    Save 9.44 €
    12% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.

SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.

  • Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
  • All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards

Specifications

Closure Size G 1 1/2 in., 32 mm
Closure Color Black
Description G 1 1/2 in. (f) to (OD) 32 mm
Color Black
Liner Material HDPE-EC
Material HDPE-EC
Product Type Thread Adapter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.