missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ HDPE-EC Safety Canister and Cap

Product Code. 17858743 Shop All SCAT Europe Products
Change view
Click to view available options
Type:
Canister
Replacement Screw Cap
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
17858743 Replacement Screw Cap Each
15745943 Canister Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17858743 Supplier SCAT™ Supplier No. 108421D
  • ONLINE EXCLUSIVE PRICE
    4.35 €
    3.96 € / Pack of 1
    Save 0.39 €
    8% Off
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Electrically conductive safety canisters made of high-quality HDPE-EC material. Due to their grounding capability, flying sparks caused by electrical discharge can be prevented when disposing of flammable or explosive laboratory liquids.

  • Made of very robust and electronically conductive HDPE, which is highly resistant to all solvents commonly used in laboratories
  • Filling quantities from 10 L for the use with HPLC systems up to massive 60 L drums for the collection of large quantities of containers (e.g., professional SymLine™ Disposal Systems)
  • Thread types S50 is commonly used in the laboratory and are compatible with SCAT™ Safety Waste Caps range for the safe disposal of volatile hazardous substances
  • Special safety features such as integrated visibility strips or floats ensure permanent fill level monitoring
  • Practical designs as space-saving canisters allow safe storage of liquid waste even in small areas or in hazardous goods cabinets
  • Ideally suited for combination with SCAT™ Safety Waste Caps or the SymLine™ Disposal Systems

Specifica

For Use With (Application) Flammable Liquid Waste Collection
For Use With (Equipment) SCAT Canister 108421
Type Replacement Screw Cap
Material HDPE
Includes Cap for SCAT™ Canister 108421
Color Black
Thread Size S50
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.