missing translation for 'onlineSavingsMsg'
Learn More

WHEATON™ WHEATON™ HDPE Liquid Scintillation Vial

Product Code. 10523804 Shop All Wheaton Science Products Products
Change view
Click to view available options
Material (Liner):
Foamed Polyethylene
Linerless
Metal foil
Quantity:
1000/Pk.
500/Pk.
Unit Size:
Pack of 1000
Pack of 500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material (Liner) Quantity Unit Size
10523804 Linerless 500/Pk. Pack of 500
11758869 Foamed Polyethylene 1000/Pk. Pack of 1000
11754179 Linerless 1000/Pk. Pack of 1000
11748839 Metal foil 1000/Pk. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10523804 Supplier WHEATON™ Supplier No. 986714
  • ONLINE EXCLUSIVE PRICE
    291.00 €
    225.39 € / Pack of 500
    Save 65.61 €
    22% Off
Estimated Shipment: 23-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

WHEATON™ HDPE Liquid Scintillation Vial is designed for reliable liquid scintillation counting. Made from high-density polyethylene with lightweight walls for increased counting efficiency, manufactured to precise tolerances, and available with a choice of cap liner materials.

  • Made from high-density polyethylene (HDPE) with lightweight walls for increased counting efficiency
  • Manufactured to precise tolerances to avoid jamming in automated equipment
  • 20 mL capacity with 22-400 cap size; dimensions 27 x 61 mm
  • Available with a choice of cap liner materials including linerless, foamed polyethylene, and metal foil options
  • Cases of 1000 vials are packed in a single plastic bag with caps supplied separately
  • Suitable for buffers, reagents, and liquid, solid, and semi-solid content

Specifications

Material (Liner) Linerless
Color Natural
Diameter (Metric) Outer 27 mm
Material HDPE
Sterility Non-sterile
Volume (Metric) 20 mL
Closure Material Polyethylene
Closure Type Screw Cap
Quantity 500/Pk.
Closure Size 22-400
Autoclavable Not Autoclavable
Height (Metric) 61 mm
Product Type Scintillation Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.