missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brand™ HDPE Scoops
Scoop powdered and granular material with ease. BRAND™ HDPE Scoops provide sturdy handling with durable polyethylene.
Brand: Brand™ 139035
Code : NEW
Additional Details : Weight : 3.84000kg
Description
- Manufactured from HDPE
- Natural colored
Specifications
Scoop | |
500 mL | |
350 mm |
HDPE | |
Natural |