missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ HDPE, Screw Closure with Tamper-evident Ring (Black)

Product Code. 15106675
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15106675 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15106675 Supplier Kautex™ Supplier No. 2000085874
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For Use With: 2/2,5/3,6/8 Jerrycan

  • Screw closure
  • HDPE
  • Tamper evident ring
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) 2/2,5/3,6/8 Jerrycan
Includes Tamper Evident Ring
Material HDPE
Product Type Screw Closure
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.