missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ HDPE, Wide Neck Jerry Can with Threaded Connector

Product Code. 10497951 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
6 L
11 L
20 L
30 L
Dimensions (L x W x H):
264 x 132 x 287 mm (D x W x H)
264 x 132 x 426 mm (D x W x H)
365 x 165 x 485 mm (D x W x H)
365 x 240 x 485 mm (D x W x H)
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Dimensions (L x W x H) Unit Size
10497951 11 L 264 x 132 x 426 mm (D x W x H) Each
10229070 6 L 264 x 132 x 287 mm (D x W x H) Each
10096771 20 L 365 x 165 x 485 mm (D x W x H) Each
10411381 30 L 365 x 240 x 485 mm (D x W x H) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10497951 Supplier Buerkle™ Supplier No. 04311010
  • ONLINE EXCLUSIVE PRICE
    71.30 €
    64.15 € / Pack of 1
    Save 7.15 €
    10% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Wide-necked canister with extremely wide filling opening (88 mm), easy to clean on the inside, which is particularly important when storing sensitive substances.

  • Wide filling opening allows for easy cleaning
  • Carrying handles and screw lid guarantee safe and convenient transport
  • Can also be used as a settling canister
  • Manufactured from transparent HDPE
  • UV stabilized
  • Self-adhesive blue litre graduation and spigots available as accessory
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Capacity (English) 2.91 gal.
Capacity (Metric) 11 L
Certifications/Compliance Suitable for foodstuffs
Closure Type Screw
Color Transparent
Dimensions (L x W x H) 264 x 132 x 426 mm (D x W x H)
For Use With (Application) Suitable for Use with Foodstuffs, Safe and Convenient Transport, Used as a Settling Canister
Graduations Liter scale available separately
Includes 0.75 in. Threaded connector, carrying handles, screw lid
Material HDPE
Mouth Style Wide
Product Type Jerry Can
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.