missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ HDPE Wide Neck Square Reagent Bottles

Product Code. 11817702 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
750 mL
1000 mL
1500 mL
2500 mL
4000 mL
Unit Size:
Each
Pack of 6
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11817702 500 mL Each
15475574 50 mL Each
15425574 100 mL Each
10222563 2500 mL Each
17250574 750 mL Each
15376418 250 mL Each
11827702 1000 mL Pack of 6
15316438 1000 mL Each
11868301 1500 mL Each
15425584 4000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 11817702 Supplier Buerkle™ Supplier No. 03420500
  • ONLINE EXCLUSIVE PRICE
    4.10 €
    3.42 € / Pack of 1
    Save 0.68 €
    16% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Wide-neck reagent bottle is particularly suited for storing, packing, taking samples, sampling and transport of granules and paste-like media.

  • HDPE, transparent
  • PP tamper-evident screw cap, blue
  • UN approval for bulk goods
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Color Transparent
Diameter (Metric) Inner Neck 43 mm
For Use With (Application) Foodstuffs
Height (Metric) With Closure 165 mm
Material HDPE, Polypropylene
Length (Metric) 75 mm
Sterility Non-sterile
Width (Metric) 75 mm
Thread Size (Metric) 54 mm
Closure Color Blue
Closure Included Yes
Graduated No
Mouth Wide
Height (Metric) 162 mm
Width (English) 2.95 in.
Length (English) 2.95 in.
Height (English) 6.38 in.
Closure Material Polypropylene
Closure Type Screw Cap With Tamper-Evident Feature
Autoclavable Not Autoclavable
Capacity (English) 16.91 oz.
Shape Square
Certifications/Compliance UN Approval
Disposable Reusable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.