missing translation for 'onlineSavingsMsg'
Learn More

THOMSON INSTRUMENT COMPANY Head Space Frits

Product Code. 30696774
Encompass_Preferred
Change view
Click to view available options
Size:
110 g/160 g
12 g
240 g/300 g
25 g
4 g
40 g
80 g, 90 g
Unit Size:
Each
Pack of 100
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30696774 4 g Pack of 100
30696718 12 g Pack of 100
10783184 25 g Each
30696705 40 g Pack of 100
30696744 80 g, 90 g Pack of 100
30696706 110 g/160 g Pack of 100
30696748 240 g/300 g Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 30696774 Supplier THOMSON INSTRUMENT COMPANY Supplier No. 491250
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use with SINGLE StEP™ Empty Column

  • SINGLE StEP™ Empty Columns designed for seamless integration with FPLC, LPLC, and MPLC chromatography systems.
  • Compatible with both gravity flow and FPLC/LPLC/MPLC applications up to 200 PSI.
  • Available in a wide range of sizes from 10 mL to 650 mL, accommodating approximately 4 g to 300 g of resin.
  • Equipped with standard Luer fittings on both the top and bottom for quick, secure connections.
  • Optional 1.2 L resin reservoir available to simplify column packing and loading.
  • The 1.2 L resin reservoir threads directly onto 25 g, 40 g, and 80 g SINGLE StEP™ empty columns.
  • Reservoir enables convenient loading of buffers, solvents, and samples while helping minimize contamination and sample loss.
  • Each column includes a bottom frit; compatible top frits are available separately.
  • Manufactured in the USA to support consistent quality and same-day shipping.

Specifications

Porosity 25 μm
Size 4 g
Type Column Frit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.