missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Heat-On Inserts

Product Code. 11731659 Shop All Heidolph Instruments Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
150 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11731659 10 mL Each
10319943 25 mL Each
10432915 150 mL Each
10617974 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11731659 Supplier Heidolph™ Supplier No. 5058006100
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Suitable for heating round bottom flasks

  • For optimal use it is recommended that you also purchase a Multi-Well holder and safety lifting handles
  • The maximum temperature for continuous use is 260°C
    • To measure this precisely with a temperature sensor, a small hole is provided on one side of the Heat-On Block
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 10 mL
For Use With (Equipment) For Multi Well Holder
Type Heat-On Insert
Voltage 100/230 V
Coating Fluoropolymer
Material Aluminium 6082 HE30
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.