Learn More
Eppendorf™ Heat Sealer S100
Product Code. 15530566 Shop All Eppendorf Products
Change view
Click to view available options
Plug Type:
EU Plug
GB Plug
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type unitSize
15530566 EU Plug Each
15775665 GB Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15530566 Supplier Eppendorf™ Supplier No. 5391000001
Only null left
Add to basket
Add to basket
Provides user-friendly and reliable hermetic sealing of a variety of multiwell plates.
  • Hermetic sealing of multiwell plates
  • Fixed sealing temperature for reproducible results
  • Eliminates evaporation in PCR, reducing cross contamination
  • Seals 96- and 384-well plates
  • Compact design, portable device
  • Integrated thermostats prevents overheating
  • Improved mechanics for even easier sealing
  • PCR plate adapter included
TRUSTED_SUSTAINABILITY
Specifications
Hertz 50/60 Hz
Voltage 100/230 V
Dimensions (L x W x H) 220 × 250 × 230 mm
Electrical Requirements 100/230 V, 50/60 Hz
For Use With (Equipment) 96- and 384-well plates
Plug Type EU Plug