missing translation for 'onlineSavingsMsg'
Learn More

Labconco Heated Product Chamber

Product Code. 17910190 Shop All Labconco Corporation Products
Change view
Click to view available options
No. of Shelves:
3
5
Includes:
Valves
Without Valves
Unit Size:
1 unit
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Shelves Includes unitSize
17910190 3 Without Valves 1 unit
17930180 5 Without Valves 1 unit
17930190 3 Valves 1 unit
17970180 5 Valves 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17910190 Supplier Labconco Supplier No. 780801090
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chambers allow for drying of bulk samples, or drying of serum bottles, vials or microplates. The 10 in. diameter shelves are wireless and removable so that they may be placed in a freezer for pre-freezing. Chamber connects to a FreeZone Freeze Dryer for control and recording of shelf temperature.

  • Chambers allow for drying of bulk samples, or drying of serum bottles, vials or microplates.
  • The 10 in. diameter shelves are wireless and removable so that they may be placed in a freezer for pre-freezing.
  • Chamber connects to a FreeZone Freeze Dryer for control and recording of shelf temperature from the Lyo-Works display.
  • Each Chamber includes a 12 in. diameter clear acrylic cylinder, vacuum release valve, cap for vacuum port and 8 in. diameter opening to collector on bottom of chamber. Will fit 10.12 in. or 14.0 in. diameter condensers.
  • Includes three shelves and three temperature probes.
  • For use with any FreeZone Freeze Dryer manufactured after July 2017.

Specifications

Type Heated Product Chamber
No. of Shelves 3
Temperature Range (Metric) -20°C to +60°C
Material Acrylic
Includes Without Valves
Plug Type All except North America
Voltage 230 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.