Learn More
Cole-Parmer™ Heating Block for Electrothermal™ RS-250D-S Shaking Reaction Stations
Shop All Cole-Parmer ProductsDescription
- Compatible with Electrothermal RS9000 Reaction Stations
Specifications
Specifications
| Description | Heating Block for Reaction System |
| For Use With (Equipment) | RS-250D-S Shaking Reaction Stations |
| Holds | 40 x 24 mm Tubes |
| No. of Places | 40 |
| Type | Heating Block |
| Warranty | 1 Year |