missing translation for 'onlineSavingsMsg'
Learn More

AZLON™ Heavy Duty HDPE Carboy with Stopcock

Product Code. 31911936
Change view
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
31911936 5 L Each
31911932 10 L Each
31911934 20 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. Se returpolicy
Produktkod. 31911936 Leverantör AZLON™ Leverantörsnummer BNH05A
  • ONLINE EXCLUSIVE PRICE
    70.49 €
    43.79 € / Pack of 1
    Save 26.70 €
    37% Off
Estimated Shipment: 08-01-2027
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Heavy‐duty HDPE carboys support safe storage and handling of media, buffers, and sensitive liquids. Manufactured from USP Class VI HDPE with a regulatory‐compliant PP cap, this product family is available with or without a leak‐resistant quarter‐turn stopcock for controlled dispensing.

  • Carboys are manufactured from USP Class VI compliant, BPA-free, FDA-grade HDPE resin for superior chemical compatibility and suitability in regulated biotech and pharmaceutical applications
  • Option of leakproof heavy duty stopcock manufactured from USP Class VI compliant PP resin for safe and easy liquid dispensing, and cap made from regulatory compliant PP
  • Reliable Performance, Every Day: From media preparation to large volume liquid handling, AZLON™ carboys deliver dependable, user friendly performance when liquid handling matters
  • Lightweight and Durable: AZLON™ carboys, made from high-quality HDPE, are durable, shatter-resistant, and lightweight - perfect for busy laboratories
  • Designed for Ease of Use and Safety: Ergonomic handles ensure a secure grip, squat form square profile provides excellent stability on the bench top or in transit whilst also optimizing valuable laboratory space, with the addition of an optional leak-proof stopcock our carboys support safe controlled dispensing
  • Practical Features That Support Your Workflow: Molded graduations enable easy volume management, caps for 10 L and 20 L carboys have molded drill guides for easy location of probes, feed lines, and fluid aspiration, and the optional leakproof heavy duty stopcock opens and closes in a quarter turn
  • Built for Compliance: USP Class VI and regulatory compliant resins ensure regulatory and operational compliance in your workflow
  • Sustainability: Product is re-usable, durable and recyclable at end of life, helping to support internal sustainability goals
  • Applications: Suitable for regulated environments such as bioprocessing, pharma, and analytical labs
  • Spare caps available to purchase separately: 45 mm Cap CAP045/H fits 5 L AZLON™ heavy duty carboy with stopcock BNH05A and 80 mm Cap CGZ85PP fits 10 L and 20 L AZLON™ heavy duty carboy with stopcock BNH10A and BNH20A

Specifikationer

Autoclavable Not Autoclavable
Capacity (Metric) 5 L
Certifications/Compliance USP Class VI Compliant, FDA‐grade, BPA‐free Materials, Regulatory‐compliant Polypropylene Components
Closure Color White
Closure Liner Linerless
Closure Material Polypropylene
Closure Size 45 mm
Color Natural
Disposable Reusable
For Use With (Application) Regulated environments such as bioprocessing, pharma, and analytical labs
Height (Metric) 270 mm
Includes Screw Cap, Stopcock, Graduations, Handle
Material HDPE
Mouth Style Narrow
Product Type Carboy
Sterility Non-sterile
Width (Metric) 220 mm
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.